missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Cytokine-inducible SH2-containing Protein
A DNA sequence encoding the Homo sapiens (Human) Cytokine-inducible SH2-containing protein, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8202-ec-50ug
Additional Details : Weight : 0.01000kg
Specifications
NP_037456.5 | |
Cytokine-inducible SH2-containing Protein | |
Research Use Only | |
CISH | |
Recombinant Protein | |
E. coli | |
MVLCVQGPRP LLAVERTGQR PLWAPSLELP KPVMQPLPAG AFLEEVAEGT PAQTESEPKV LDPEEDLLCI AKTFSYLRES GWYWGSITAS EARQHLQKMP EGTFLVRDST HPSYLFTLSV KTTRGPTNVR IEYADSSFRL DSNCLSRPRI LAFPDVVSLV QHYVASCTAD TRSDSPDPAP TPALPMPKED APSDPALPAP PPATAVHLKL VQPFVRRSSA RSLQHLCRLV INRLVADVDC LPLPRRMADY LRQYPFQL | |
Greater than 80% as determined by SDS-PAGE. |
1154 | |
50 μg | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |