missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Cytochrome c1, heme protein, mitochondrial Protein

A DNA sequence encoding the Homo sapiens (Human) Cytochrome c1, heme protein, mitochondrial, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP8372-ec-10ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16084271

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

Specifications

Specifications

NP_001907.2
Cytochrome c1, heme protein, mitochondrial Protein
Research Use Only
CYC1
Recombinant Protein
E. coli
SDLELHPPSY PWSHRGLLSS LDHTSIRRGF QVYKQVCASC HSMDFVAYRH LVGVCYTEDE AKELAAEVEV QDGPNEDGEM FMRPGKLFDY FPKPYPNSEA ARAANNGALP PDLSYIVRAR HGGEDYVFSL LTGYCEPPTG VSLREGLYFN PYFPGQAIAM APPIYTDVLE FDDGTPATMS QIAKDVCTFL RWASEPEHDH RKRMGLKMLM MMALLVPLVY TIKRHKWSVL KSRKLAYRPP K
Greater than 90% as determined by SDS-PAGE.
1537
10 μg
Not determined.
Full Length
Human
GST
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers