missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Cytochrome C Protein
A DNA sequence encoding the Homo sapiens (Human) Cytochrome C, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP5900-ec-50ug
Additional Details : Weight : 0.01000kg
Specifications
NP_061820.1 | |
Cytochrome C Protein | |
Research Use Only | |
CYCS | |
Recombinant Protein | |
E. coli | |
GDVEKGKKIF IMKCSQCHTV EKGGKHKTGP NLHGLFGRKT GQAPGYSYTA ANKNKGIIWG EDTLMEYLEN PKKYIPGTKM IFVGIKKKEE RADLIAYLKK ATNE | |
Greater than 90% as determined by SDS-PAGE. |
54205 | |
50 μg | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |