missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Recombinant Human Cystatin C Protein

Product Code. 18677207 Shop All Bio Techne Products
Click to view available options
Quantity:
1 mg
100 μg
5 μg
500 μg
Unit Size:
100µg
1mg
500µg
5µg
This item is not returnable. View return policy

Product Code. 18677207

Brand: Novus Biologicals™ NBP2613815ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Data Not Available.
TRUSTED_SUSTAINABILITY

Specifications

For Use With (Application) Western Blot
Formulation 0.2 μm filtered solution in 20 mM Tris-HCl, pH 8.0, 300 mM NaCl
Gene ID (Entrez) 1471
Molecular Weight (g/mol) MolecularWeight-theroretical: 13.3 kDa
Quantity 5 μg
Storage Requirements Store at -20°C. Avoid freeze/thaw cycles.
Endotoxin Concentration Less than 0.1 EU/μg of Cystatin C as determined by LAL method.
Gene Alias ARMD11, bA218C14.4 (cystatin C), cystatin 3, cystatin C, cystatin C (amyloid angiopathy and cerebral hemorrhage), cystatin-3, cystatin-C, Gamma-trace, MGC117328, Neuroendocrine basic polypeptide, post-gamma-globulin
Gene Symbol CST3
Research Category Cancer, Epitope Tags, Signal Transduction
Purity or Quality Grade >98%, by SDS-PAGE and HPLC
Protein Cystatin C
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.