missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Recombinant Human CYP4F3 GST (N-Term) Protein

Product Code. 18326797 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
10 μg
25 μg
Unit Size:
10µg
25µg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18326797 25 μg 25µg
18356687 10 μg 10µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Este artículo no se puede devolver. Vea la política de devoluciones
Código de producto. 18326797 Proveedor Novus Biologicals™ N.º de proveedor H00004051Q0125ug

Please to purchase this item. Need a web account? Register with us today!

Este artículo no se puede devolver. Vea la política de devoluciones

Recombinant Protein

This protein was produced in an in vitro wheat germ expression system that should preserve correct conformational folding that is necessary for biological function. While it is possible that this protein could display some level of activity, the functionality of this protein has not been explicitly measured or validated.
TRUSTED_SUSTAINABILITY

Especificaciones

For Use With (Application) Western Blot, ELISA, PAGE, Protein Array, Immunoaffinity Purification
Formulation 50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer.
Gene ID (Entrez) 4051
Molecular Weight (g/mol) 36.63 kDa
Quantity 25 μg
Storage Requirements Store at -80°C. Avoid freeze-thaw cycles.
Gene Alias CYP4F, CYPIVF3, cytochrome P-450, Cytochrome P450 4F3, cytochrome P450, family 4, subfamily F, polypeptide 3, cytochrome P450, subfamily IVF, polypeptide 3 (leukotriene B4 omegahydroxylase), Cytochrome P450-LTB-omega, EC 1.14.13.30, leukotriene B4 omega hydroxylase, Leukotriene-B(4) 20-monooxygenase 2, leukotriene-B(4) omega-hydroxylase 2, leukotriene-B4 20-monooxygenase, LTB4HCPF3
Gene Symbol CYP4F3
Research Category Lipid and Metabolism
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Protein CYP4F3
Mostrar más Mostrar menos
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.