missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Cyclophilin G Protein

Product Code. 15984288
Click to view available options
Quantity:
1 mg
25 μg
5 μg
Unit Size:
1mg
25µg
5µg
This item is not returnable. View return policy

Product Code. 15984288

Brand: enQuireBio™ QP105741mg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

A cDNA sequence encoding the Cyclophilin G was constructed and used to recombinantly synthesize the protein.

Specifications

Gene ID (Entrez) 9360
Name Cyclophilin G Protein
Quantity 1 mg
Regulatory Status Research Use Only
Endotoxin Concentration < 1.0 EU per ug protein as determined by the LAL method.
Gene Symbol PPIG
Biological Activity Specific activity is > 200 nmoles/min/mg, and is defined as the amount of enzyme that cleaves 1umole of suc-AAFP-pNA per minute at 25C in Tris-Hcl pH8.0 using chymotrypsin.
Product Type Recombinant Protein
Cross Reactivity Human
Species E. coli
Protein Tag His
Sequence MGSSHHHHHH SSGLVPRGSH MGIKVQRPRC FFDIAINNQP AGRVVFELFS DVCPKTCENF RCLCTGEKGT GKSTQKPLHY KSCLFHRVVK DFMVQGGDFS EGNGRGGESI YGGFFEDESF AVKHNKEFLL SMANRGKDTN GSQFFITTKP TPHLDGHHVV FGQVISGQEV VREIENQKTD AASKPFAEVR ILSCG
Buffer Cyclophilin-G solution containing 20mM Tris-HCl pH 8.0, 1mM DTT and 10% glycerol.
Purity or Quality Grade Greater than 95.0% as determined by SDS-PAGE.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.