missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Recombinant Human Cyclophilin-F Protein

Product Code. 18675256 Shop All Bio Techne Products
Click to view available options
Quantity:
10 μg
500 μg
Unit Size:
10µg
500µg
This item is not returnable. View return policy

Product Code. 18675256

Brand: Novus Biologicals™ NBP26136910ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Data Not Available.
TRUSTED_SUSTAINABILITY

Specifications

Formulation 0.2 μm filtered solution in PBS, pH 7.4, 1 mM DTT, 10 % glycerol.
Gene ID (Entrez) 10105
Molecular Weight (g/mol) MolecularWeight-theroretical: 18.9 kDa
Quantity 10 μg
Storage Requirements Store at -20°C. Avoid freeze/thaw cycles.
Endotoxin Concentration Less than 0.1 EU/μg of Cyclophilin-F as determined by LAL method.
Gene Alias Cyclophilin F, CYP3FLJ90798, Cyp-D, hCyP3, MGC117207, mitochondrial, peptidyl-prolyl cis-trans isomerase, mitochondrial, peptidylprolyl isomerase F, peptidylprolyl isomerase F (cyclophilin F)
Gene Symbol PPIF
Research Category Proteases & Other Enzymes
Purity or Quality Grade >95%, by SDS-PAGE and HPLC
Protein Cyclophilin-F
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.