missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Cyclophilin B Protein
A cDNA sequence encoding the Cyclophilin B was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP10570-50ug
Additional Details : Weight : 0.01000kg
Specifications
5479 | |
50 μg | |
< 1.0 EU per ug protein as determined by the LAL method. | |
Specific activity is > 220 nmoles/min/mg, and is defined as the amount of enzyme that cleaves 1umole of suc-AAFP-pNA per minute at 25C in Tris-Hcl pH8.0 using chymotrypsin. | |
Human | |
Untagged | |
1 mg/ml solution containing 20mM Tris-HCl 8.0, 20mM NaCl, 0.5mM DTT and 10% glycerol. |
Cyclophilin B Protein | |
Research Use Only | |
PPIB | |
Recombinant Protein | |
E. coli | |
MLLPGPSAAD EKKKGPKVTV KVYFDLRIGD EDVGRVIFGL FGKTVPKTVD NFVALATGEKGFGYKNSKFH RVIKDFMIQG GDFTRGDGTG GKSIYGERFP DENFKLKHYG PGWVSMANAGKDTNGSQFFI TTVKTAWLDG KHVVFGKVLE GMEVVRKVES TKTDSRDKPL KDVIIADCGK IEVEKPFAIA KE | |
Greater than 95.0% as determined by SDS-PAGE. |