missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human CYB5R2 Protein
A cDNA sequence encoding the CYB5R2 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP11574-20ug
Additional Details : Weight : 0.01000kg
Specifications
CYB5R2 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMNSRRRE PITLQDPEAK YPLPLIEKEK ISHNTRRFRF GLPSPDHVLG LPVGNYVQLL AKIDNELVVR AYTPVSSDDD RGFVDLIIKI YFKNVHPQYP EGGKMTQYLE NMKIGETIFF RGPRGRLFYH GPGNLGIRPD QTSEPKKTLA DHLGMIAGGT GITPMLQLIR HITKDPSDRT RMSLIFANQT EEDILVRKEL EEIARTHPDQ FNLWYTLDRP PIGWKYSSGF VTADMIKEHL PPPAKSTLIL VCGPPPLIQT AAHPNLEKLG YTQDMIFTY | |
Greater than 90.0% as determined by SDS-PAGE. |
20 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
CYB5R2 protein solution (1 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 0.15M NaCl, 10% glycerol and 1mM DTT. |