missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Recombinant Human CXCL8/IL-8 Protein

Product Code. 18417431 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
1 mg
100 μg
5 μg
500 μg
Unit Size:
100µg
1mg
500µg
5µg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18417431 5 μg 5µg
18407701 1 mg 1mg
18446011 100 μg 100µg
18442472 500 μg 500µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 18417431 Supplier Novus Biologicals™ Supplier No. NBP2349055ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Fully biologically active when compared to standard. The ED50 as determined by a chemotaxis bioassay using human CXCR2 transfected mouse BaF3 cells is less than 2 ng/mL, corresponding to a specific activity of >5.0 x 10^5 IU/mg.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P10145, P10145, P10145
Concentration LYOPH
For Use With (Application) PAGE, Bioactivity
Formulation Lyophilized from a 0.2 um filtered concentrated solution in PBS, pH 7.4.
Gene ID (Entrez) 3576
Molecular Weight (g/mol) 8.4 kDa
Quantity 5 μg
Storage Requirements Store at -20°C to -70°C as supplied. After reconstitution, store at 2°C to 8°C for 1 month and at -20°C to -70°C for long term storage. Avoid repeated freeze-thaw cycles.
Endotoxin Concentration Less than 1 EU/ug of CXCL8/IL-8 as determined by LAL method.
Gene Alias 3-10C, AMCF-I, C-X-C motif chemokine 8, CXCL8SCYB8, Emoctakin, GCP-1TSG-1, IL-8/CXCL8, interleukin 8, K60, LECT, MDNCFb-ENAP, member 8, MONAPGCP1, NAP-1NAP1, Neutrophil-activating protein 1, Protein 3-10C, T cell chemotactic factor, T-cell chemotactic factor
Gene Symbol CXCL8
Reconstitution Recommended to centrifuge prior to opening. Reconstitute in sterile distilled water or aqueous buffer containing 0.1% BSA to a concentration of 0.1-1.0mg/mL. Apportion stock solutions into working aliquots and store at <-20°C.
Research Category Angiogenesis, Biologically Active Proteins, Cancer, Cytokine Research, Immunology, Innate Immunity, Signal Transduction
Purity or Quality Grade >97% pure by SDS-PAGE and HPLC
Protein CXCL8/IL-8
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.