missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Recombinant Human CXCL3/GRO gamma/CINC-2/DCIP-1 Protein
Shop All Bio Techne ProductsDescription
Fully biologically active when compared to standard. The biological activity determined by a chemotaxis bioassay using human CXCR2 transfected human 293 cells is in a concentration range of 10-100 ng/mL.
Specifications
Specifications
| Accession Number | P19876, P19876, P19876 |
| Concentration | LYOPH |
| For Use With (Application) | PAGE, Bioactivity |
| Formulation | Lyophilized from a 0.2 um filtered concentrated solution in 20 mM PB, pH 7.4, 50 mM NaCl. |
| Gene ID (Entrez) | 2921 |
| Molecular Weight (g/mol) | 7.9 kDa |
| Quantity | 1 mg |
| Storage Requirements | Store at -20°C to -70°C as supplied. After reconstitution, store at 2°C to 8°C for 1 month and at -20°C to -70°C for long term storage. Avoid repeated freeze-thaw cycles. |
| Endotoxin Concentration | Less than 1 EU/ug of CXCL3/GRO gamma/CINC-2/DCIP-1 as determined by LAL method. |
| Gene Alias | chemokine (C-X-C motif) ligand 3, CINC-2 alpha, CINC-2 alpha/CXCL3, CINC-2 beta, CINC-2 beta/CXCL3, CINC-2b, C-X-C motif chemokine 3, DCIP-1, DCIP-1/CXCL3, GRO3, GRO3 oncogene, GROg, GRO-gamma, GRO-gamma(1-73), Growth-regulated protein gamma, Macrophage inflammatory protein 2-beta, melanoma growth stimulatory activity gamma, MGSA gamma, MIP-2b, MIP2B, MIP2-beta, SCYB3 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?