missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Recombinant Human CXCL13/BLC/BCA-1 Protein

Product Code. 18430432 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
1 mg
100 μg
5 μg
500 μg
Unit Size:
100µg
1mg
500µg
5µg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18430432 500 μg 500µg
18403812 5 μg 5µg
18414352 1 mg 1mg
18418591 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 18430432 Supplier Novus Biologicals™ Supplier No. NBP234885500ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

CXCL13 protein is fully biologically active when compared to standard. The biological activity determined by a chemotaxis bioassay using human B cells is in a concentration range of 1.0-10 ng/mL.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number O43927, O43927, O43927
Concentration LYOPH
For Use With (Application) PAGE, Bioactivity
Formulation Lyophilized from a 0.2 um filtered concentrated solution in 20 mM PB, pH 7.4, 100 mM NaCl.
Gene ID (Entrez) 10563
Molecular Weight (g/mol) 10.3 kDa
Quantity 500 μg
Storage Requirements Store at -20°C to -70°C as supplied. After reconstitution, store at 2°C to 8°C for 1 month and at -20°C to -70°C for long term storage. Avoid repeated freeze-thaw cycles.
Endotoxin Concentration Less than 1 EU/ug of CXCL13/BLC/BCA-1 as determined by LAL method.
Gene Alias ANGIE, ANGIE2, B cell-attracting chemokine 1, B lymphocyte chemoattractant, BCA-1/CXCL13, BCA1B-cell chemoattractant, BCA-1CXC chemokine BLC, B-cell-homing chemokine (ligand for Burkitt's lymphoma receptor-1), BLCSmall-inducible cytokine B13, BLR1L, B-lymphocyte chemoattractant, chemokine (C-X-C motif) ligand 13, chemokine (C-X-C motif) ligand 13 (B-cell chemoattractant), C-X-C motif chemokine 13, SCYB13B-cell-attracting chemokine 1, small inducible cytokine B subfamily (Cys-X-Cys motif), member 13 (B-cellchemoattractant)
Gene Symbol CXCL13
Reconstitution Recommended to centrifuge prior to opening. Reconstitute in sterile distilled water or aqueous buffer containing 0.1% BSA to a concentration of 0.1-1.0mg/mL. Apportion stock solutions into working aliquots and store at <-20°C.
Research Category Biologically Active Proteins, Chemokines and Cytokines
Purity or Quality Grade >97% pure by SDS-PAGE and HPLC
Protein CXCL13/BLC/BCA-1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.