missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Recombinant Human CXCL12/SDF-1 alpha Protein
Shop All Bio Techne Products
Click to view available options
Quantity:
10 μg
100 μg
1000 μg
500 μg
Unit Size:
1000µg
100µg
10µg
500µg
Description
SDF1 alpha Protein is fully biologically active when compared to standard. The biological activity determined by a chemotaxis bioassay using PHA and rHuIL-2 activated human peripheral blood T-lymphocytes is in a concentration range of 20-80 ng/mL.
Specifications
Specifications
| Accession Number | P48061, P48061, P48061 |
| For Use With (Application) | SDS-PAGE, Bioactivity |
| Formulation | Lyophilized from a 0.2 μm filtered concentrated solution in 20 mM PB pH 7.0, 130 mM NaCl. |
| Gene ID (Entrez) | 6387 |
| Molecular Weight (g/mol) | MolecularWeight-theroretical: 8.0 kDa |
| Quantity | 500 μg |
| Storage Requirements | Store at -20°C to -70°C as supplied. After reconstitution, store at 2 to 8 C for 1 month and (at -20°C to -70°C for long term storage. Avoid repeated freeze/thaw cycles.) |
| Endotoxin Concentration | Less than 1 EU/μg of CXCL12/SDF-1 alpha as determined by LAL method. |
| Gene Alias | CXCL12 chemokine (C-X-C motif) ligand 12, IRH, PBSF, SCYB12, SDF1, SDF-1 alpha, SDF-1 alpha/CXCL12 alpha, SDF1A, SDF1B, TLSF, TPAR1 |
| Gene Symbol | CXCL12 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction