missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Human CXCL10/IP-10 Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity
£160.90 - £408.20
Specifications
Gene ID (Entrez) | 3627 |
---|---|
Molecular Weight (g/mol) | 8.7 kDa |
Name | CXCL10/IP-10/CRG-2 |
Source | E. coli-derived human CXCL10/IP-10/CRG-2 protein Val22-Pro98,with an N-terminal Met |
Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. |
Product Code | Brand | Formulation | Quantity | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Formulation | Quantity | Price | Quantity & Availability | ||||
18320088
|
R&D Systems™
266-IP-010 |
£160.90
10µg |
Estimated Shipment: 21-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18319267
|
R&D Systems™
266-IP-050 |
£408.20
50µg |
Estimated Shipment: 21-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18118660
|
R&D Systems™
266-IP-050/CF |
Lyophilized from a 0.2μm filtered solution in PBS. | 50μg |
£408.20
50µg |
Estimated Shipment: 21-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
18330288
|
R&D Systems™
266-IP-010/CF |
£160.90
10µg |
Estimated Shipment: 21-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The Recombinant Human CXCL10/IP-10 Protein is derived from E. coli. The Recombinant Human CXCL10/IP-10 Protein has been validated for the following applications: Bioactivity.Specifications
3627 | |
CXCL10/IP-10/CRG-2 | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. | |
Recombinant |
8.7 kDa | |
E. coli-derived human CXCL10/IP-10/CRG-2 protein Val22-Pro98,with an N-terminal Met | |
Unconjugated | |
97%, by SDS-PAGE under reducing conditions and visualized by silver stain. |