missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human CUTA Protein
A cDNA sequence encoding the CUTA was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP11568-1mg
Additional Details : Weight : 0.01000kg
Specifications
CUTA Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MRLLLLPRVL LTMASGSPPT QPSPASDSGS GYVPGSVSAA FVTCPNEKVA KEIARAVVEK RLAACVNLIP QITSIYEWKG KIEEDSEVLM MIKTQSSLVP ALTDFVRSVH PYEVAEVIAL PVEQGNFPYL QWVRQVTESV SDSITVLPLE HHHHHH | |
Greater than 95% as determined by SDS-PAGE. |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
CUTA Human solution containing 20mM Tris-HCl pH 8.0, 1mM DTT, and 10% glycerol. |