missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Human CUL2/RBX1 Complex Protein

Product Code. 18748376
Change view
Click to view available options
Quantity:
25 μg
Unit Size:
25µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18748376 25 μg 25µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18748376 Supplier R&D Systems™ Supplier No. E3420025
 Restricted Item
Estimated Shipment: 03-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Typical enzyme conentration to support conjugation in vitro will depend on experimental conditions.

Typical enzyme concentration to support in vitro conjugation will depend on experimental conditions.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q13617.2 (CUL2), P62877.1 (RBX1)
For Use With (Application) Bioactivity
Formulation Supplied as a 0.2 μm filtered solution in HEPES, NaCl, DTT and Glycerol.
Molecular Weight (g/mol) M.W. (theoretical): 93 kDa (CUL2), 12 kDa (RB x 1)
Quantity 25 μg
Source Spodoptera frugiperda, Sf 21 (baculovirus)-derived human CUL2/RB x 1 Complex protein Met1 - Ala 745 with a N-terminal 10-His Sumo tag (CUL2), Met1 - His108 (RB x 1)
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -70°C as supplied. 3 months, -70°C under sterile conditions after opening.
Conjugate Unconjugated
Species Human
Buffer X mg/mL (X μm) in 50 mm HEPES pH 7.5, 200 mm NaCl, 10% (v/v) Glycerol, 1 mm DTT
Product Line R&D Systems
Purity or Quality Grade >90%, by SDS-PAGE under reducing conditions and visualized by Colloidal Coomassie™ Blue stain.
Protein CUL2/RBX1 Complex
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.