missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human CUE domain-containing protein 1
A DNA sequence encoding the Homo sapiens (Human) CUE domain-containing protein 2, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8197-ec-10ug
Additional Details : Weight : 0.01000kg
Specifications
NP_076945.2 | |
CUE domain-containing protein 1 | |
Research Use Only | |
CUEDC2 | |
Recombinant Protein | |
E. coli | |
MELERIVSAA LLAFVQTHLP EADLSGLDEV IFSYVLGVLE DLGPSGPSEE NFDMEAFTEM MEAYVPGFAH IPRGTIGDMM QKLSGQLSDA RNKENLQPQS SGVQGQVPIS PEPLQRPEML KEETRSSAAA AADTQDEATG AEEELLPGVD VLLEVFPTCS VEQAQWVLAK ARGDLEEAVQ MLVEGKEEGP AAWEGPNQDL PRRLRGPQKD ELKSFILQKY MMVDSA | |
Greater than 90% as determined by SDS-PAGE. |
79004 | |
10 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |