missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human CTGF Protein

Product Code. p-7152084
Click to view available options
Quantity:
1 mg
100 μg
20 μg
5 μg
500 μg
Unit Size:
100µg
1mg
20µg
500µg
5µg
This item is not returnable. View return policy

Product Code. 15946009

Brand: enQuireBio™ QP53735ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Specifications

Accession Number NP_001892.1
Gene ID (Entrez) 1490
Name CTGF Protein
Quantity 5 μg
Regulatory Status Research Use Only
Endotoxin Concentration Less than or equal to 1.00 EUs/ug as determined by the LAL method.
Gene Symbol CTGF
Biological Activity Bioactivity of Human CTGF confirmed in HUVEC cell proliferation assay.
Product Type Recombinant Protein
Cross Reactivity Human
Species E. coli
Protein Tag Untagged
Sequence MGKKCIRTP K. ISKPIKFE LS. GCTSMKT YRA. KFCGVC TDGR. CCTPH RTTTL. PVEF KCPDGE. VMK KNMMFIK. TC ACHYNCPG. D NDIFESLYY.  RKMYGDMA
Buffer Sterile filtered (0.22 um) solution containing 0.1% TFA
Purity or Quality Grade Greater than 90 as determined by SDS-PAGE.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.