missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human CTF1 Protein

A cDNA sequence encoding the CTF1 was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP11552-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15947618

  • £5160.00 / 1mg
Estimated Shipment: 13-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

1489
1 mg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Recombinant Protein
E. coli
MRGSHHHHHH GSSRREGSLE DPQTDSSVSL LPHLEAKIRQ THSLAHLLTK YAEQLLQEYV QLQGDPFGLPSFSPPRLPVA GLSAPAPSHA GLPVHERLRL DAAALAALPP LLDAVCRRQA ELNPRAPRLL RRLEDAARQA RALGAAVEAL LAALGAANRG PRAEPPAATA SAASATGVFP AKVLGLRVCG LYREWLSRTE GDLGQLLPGG SA
Purity of CTF1 Human Recombinant is greater than 90% as determined by SDS-PAGE.
CTF1 Protein
Research Use Only
CTF1
Human
His
CTF1 was filtered (0.4microm) and lyophilized from 0.5 mg/ml in 0.05M Acetate buffer pH 4.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers