missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human CSNK2B Protein
A cDNA sequence encoding the CSNK2B was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP11534-10ug
Additional Details : Weight : 0.01000kg
Specifications
1460 | |
10 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
MSSSEEVSWI SWFCGLRGNE FFCEVDEDYI QDKFNLTGLN EQVPHYRQAL DMILDLEPDE ELEDNPNQSD LIEQAAEMLY GLIHARYILT NRGIAQMLEK YQQGDFGYCP RVYCENQPML PIGLSDIPGE AMVKLYCPKC MDVYTPKSSR HHHTDGAYFG TGFPHMLFMV HPEYRPKRPA NQFVPRLYGF KIHPMAYQLQ LQAASNFKSP VKTIR | |
Greater than 95.0% as determinedAnalysis by SDS-PAGE. |
CSNK2B Protein | |
Research Use Only | |
CSNK2B | |
Human | |
Untagged | |
The CSNK2B protein (1 mg/ml) contains 20mM Tris-HCl pH 8.0, 200mM NaCl, 1mM DTT, 1mM EDTA, 1uM leupeptin and 40% glycerol. |