missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human CRYGC Protein

Product Code. 15947518
Click to view available options
Quantity:
1 mg
20 μg
5 μg
Unit Size:
1mg
20µg
5µg
This item is not returnable. View return policy

Product Code. 15947518

Brand: enQuireBio™ QP115215ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

A cDNA sequence encoding the CRYGC was constructed and used to recombinantly synthesize the protein.

Specifications

Name CRYGC Protein
Quantity 5 μg
Regulatory Status Research Use Only
Endotoxin Concentration Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Product Type Recombinant Protein
Cross Reactivity Human
Species E. coli
Protein Tag His
Sequence MGSSHHHHHH SSGLVPRGSH MGSHMGKITF YEDRAFQGRS YETTTDCPNL QPYFSRCNSI RVESGCWMLY ERPNYQGQQY LLRRGEYPDY QQWMGLSDSI RSCCLIPQTV SHRLRLYERE DHKGLMMELS EDCPSIQDRF HLSEIRSLHV LEGCWVLYEL PNYRGRQYLL RPQEYRRCQD WGAMDAKAGS LRRVVDLY
Buffer The CRYGC solution (1 mg/1ml) contains 20mM Tris-HCl buffer (pH 8.0), 200mM NaCl, 2mM DTT and 10% glycerol.
Purity or Quality Grade Greater than 95% as determined by SDS-PAGE.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.