missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human CRISP1 Protein

A cDNA sequence encoding the CRISP1 was constructed and used to recombinantly synthesize the protein.

£98.39 - £5741.17

Specifications

Name CRISP1 Protein
Product Type Recombinant Protein
Protein Tag His
Host Species E. coli
Cross Reactivity Human
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15957468
View Documents Promotion Details
enQuireBio™
QP11505-2UG
2 μg
£98.39
2µg
View alternative products
Save Up to 
»
Estimated Shipment: 01-03-2022
Please to purchase this item. Need a web account? Register with us today!
15937468
View Documents Promotion Details
enQuireBio™
QP11505-10UG
10 μg
£151.37
10µg
View alternative products
Save Up to 
»
Estimated Shipment: 01-03-2022
Please to purchase this item. Need a web account? Register with us today!
15947468
View Documents Promotion Details
enQuireBio™
QP11505-1MG
1 mg
£5741.17
1mg
View alternative products
Save Up to 
»
Estimated Shipment: 01-03-2022
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Specifications

CRISP1 Protein
His
Human
Greater than 85% as determined by SDS-PAGE.
Recombinant Protein
E. coli
MKHHHHHHAS KKKSARDQFN KLVTDLPNVQ EEIVNIHNAL RRRVVPPASN MLKMSWSEEA AQNARIFSKY CDMTESNPLE RRLPNTFCGE NMHMTSYPVS WSSVIGVWYS ESTSFKHGEW TTTDDDITTD HYTQIVWATS YLIGCAIASC RQQGSPRYLY VCHYCHEGND PETKNEPYKT GVPCEACPSN CEDKLCTNPC IYYDEYFDCD IQVHYLGCNH STTILFCKAT CLCDTEIK
SDS
Product Certifications