missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human CRHBP Protein
Click to view available options
Quantity:
1 mg
10 μg
2 μg
Unit Size:
10µg
1mg
2µg
Specifications
Specifications
| Name | CRHBP Protein |
| Quantity | 10 μg |
| Regulatory Status | Research Use Only |
| Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
| Product Type | Recombinant Protein |
| Cross Reactivity | Human |
| Species | E. coli |
| Protein Tag | His |
| Sequence | MKHHHHHHAS YLELREAADY DPFLLFSANL KRELAGEQPY RRALRCLDML SLQGQFTFTA DRPQLHCAAF FISEPEEFIT IHYDQVSIDC QGGDFLKVFD GWILKGEKFP SSQDHPLPSA ERYIDFCESG LSRRSIRSSQ NVAMIFFRVH EPGNGFTLTI KTDPNLFPCN VISQTPNGKF TLVVPHQHRN CSFSIIYPVV IKISDLTLGH VNGLQLKKSS AGCEGIGDFV ELLGGTGLDP SKMTPLADLC YPFHGPAQMK VGCDNTVVRM VSSGKHVNRV TFEYRQLEPY ELENPNGNSI GEFCLSGL YLELREAADY DPFLLFSANL KRELAGEQPY RRALRCLDML SLQGQFTFTA DRPQLHCAAF FISEPEEFIT IHYDQVSIDC QGGDFLKVFD GWILKGEKFP SSQDHPLPSA ERYIDFCESG LSRRSIRSSQ NVAMIFFRVH EPGNGFTLTI KTDPNLFPCN VISQTPNGKF TLVVPHQHRN CSFSIIYPVV IKISDLTLGH VNGLQLKKSS AGCEGIGDFV ELLGGTGLDP SKMTPLADLC YPFHGPAQMK VGCDNTVVRM VSSGKHVNRV TFEYRQLEPY ELENPNGNSI GEFCLSGL |
| Buffer | CRHBP Human was filtered (0.4microm) and lyophilized from 0.5 mg/ml supplied in 20mM TRIS and 20mM NaCl, pH 7.5. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction