missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human CREB1 Protein

A DNA sequence encoding the Homo sapiens (Human) CREB1, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP5866-ec-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15938319

  • £1735.00 / 1mg
Estimated Shipment: 12-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_004370.1
CREB1 Protein
Research Use Only
CREB1
Recombinant Protein
E. coli
MTMESGAENQ QSGDAAVTEA ENQQMTVQAQ PQIATLAQVS MPAAHATSSA PTVTLVQLPN GQTVQVHGVI QAAQPSVIQS PQVQTVQSSC KDLKRLFSGT QISTIAESED SQESVDSVTD SQKRREILSR RPSYRKILND LSSDAPGVPR IEEEKSEEET SAPAITTVTV PTPIYQTSSG QYIAITQGGA IQLANNGTDG VQGLQTLTMT NAAATQPGTT ILQYAQTTDG QQILVPSNQV VVQAASGDVQ TYQIRTAPTS TIAPGVVMAS SPALPTQPAE EAARKREVRL MKNREAAREC RRKKKEYVKC LENRVAVLEN QNKTLIEELK ALKDLYCHKS D
Greater than 90% as determined by SDS-PAGE.
1385
1 mg
Not determined.
Full Length
Human
His
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers