missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human CPOX Protein
A cDNA sequence encoding the CPOX was constructed and used to recombinantly synthesize the protein.
£99.15 - £2975.00
Specifications
Name | CPOX Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Product Type | Recombinant Protein |
Cross Reactivity | Human |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15987438
|
enQuireBio™
QP11496-5UG |
5 μg |
£99.15
5µg |
Estimated Shipment: 19-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15977438
|
enQuireBio™
QP11496-20UG |
20 μg |
£155.00
20µg |
Estimated Shipment: 19-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15967438
|
enQuireBio™
QP11496-1MG |
1 mg |
£2975.00
1mg |
Estimated Shipment: 19-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
CPOX Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
The CPOX solution (1 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 0.15M NaCl, 10% glycerol and 1mM DTT. |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSTSLGRPE EEEDELAHRC SSFMAPPVTD LGELRRRPGD MKTKMELLIL ETQAQVCQAL AQVDGGANFS VDRWERKEGG GGISCVLQDG CVFEKAGVSI SVVHGNLSEE AAKQMRSRGK VLKTKDGKLP FCAMGVSSVI HPKNPHAPTI HFNYRYFEVE EADGNKQWWF GGGCDLTPTY LNQEDAVHFH RTLKEACDQH GPDLYPKFKK WCDDYFFIAH RGERRGIGGI FFDDLDSPSK EEVFRFVQSC ARAVVPSYIP LVKKHCDDSF TPQEKLWQQL RRGRYVEFNL LYDRGTKFGL FTPGSRIESI LMSLPLTARW EYMHSPSENS KEAEILEVLR HPRDWVR | |
Greater than 95% as determined by SDS-PAGE. |