missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human CPLX1 Protein
A cDNA sequence encoding the CPLX1 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP11495-1mg
Additional Details : Weight : 0.01000kg
Specifications
CPLX1 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MEFVMKQALG GATKDMGKML GGDEEKDPDA AKKEEERQEA LRQAEEERKA KYAKMEAERE AVRQGIRDKYGIKKKEEREA EAQAAMEANS EGSLTRPKKA IPPGCGDEVE EEDESILDTV IKYLPGPLQD MLKK | |
Greater than 90% by SDS-PAGE. |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
The CPLX1protein solution contains 20mM Tris-HCl pH 8 and 10% glycerol. |