missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Corticotropin-releasing factor receptor 1 Protein

A DNA sequence encoding the Homo sapiens (Human) Corticotropin-releasing factor receptor 1, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP9080-ye-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16077581

  • £467.00 / 100µg
Estimated Shipment: 16-01-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_001138618.1
Corticotropin-releasing factor receptor 1 Protein
Research Use Only
CRHR1
Recombinant Protein
Yeast
ASLQDQHCES LSLASNISGL QCNASVDLIG TCWPRSPAGQ LVVRPCPAFF YGVRYNTTNN GYRECLANGS WAARVNYSEC QEILNEEKKS KVHYHVAVI
Greater than 90% as determined by SDS-PAGE.
104909134
100 μg
Not determined.
Partial (see sequence information for more details).
Human
His
Tris-based buffer, 50% glycerol
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers