missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human COP9 signalosome complex subunit 7a Protein
A DNA sequence encoding the Homo sapiens (Human) COP9 signalosome complex subunit 7a, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8160-ec-200ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001157565.1 | |
COP9 signalosome complex subunit 7a Protein | |
Research Use Only | |
COPS7A | |
Recombinant Protein | |
E. coli | |
SAEVKVTGQN QEQFLLLAKS AKGAALATLI HQVLEAPGVY VFGELLDMPN VRELAESDFA STFRLLTVFA YGTYADYLAE ARNLPPLTEA QKNKLRHLSV VTLAAKVKCI PYAVLLEALA LRNVRQLEDL VIEAVYADVL RGSLDQRNQR LEVDYSIGRD IQRQDLSAIA RTLQEWCVGC EVVLSGIEEQ VSRANQHKEQ QLGLKQQIES EVANLKKTIK VTTAAAAAAT SQDPEQHLTE LREPAPGTNQ RQPSKKASKG KGLRGSAKIW SKSN | |
Greater than 90% as determined by SDS-PAGE. |
50813 | |
200 μg | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |