missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Human Contactin-1 Fc Chimera Protein
Description
Measured by its binding ability in a functional ELISA. Immobilized Recombinant Human (rh) NrCAM Fc Chimera (Catalog # 2034-NR ) at 1 μg/mL (100 μL/well) can bind rhContactin-1 Fc Chimera with a linear range of 0.3-20 ng/mL. Measured by its ability to support the adhesion of C6 rat glioma cells. rhContactin Fc Chimera immobilized at 2 μg/mL, 100 μL/well will mediate >20% C6 cell adhesion.
Specifications
Specifications
| Formulation | Lyophilized from a 0.2μm filtered solution in PBS. |
| Gene ID (Entrez) | 1272 |
| Molecular Weight (g/mol) | 135 kDa |
| Name | Contactin-1 Fc Chimera |
| Quantity | 100 μg |
| Source | Spodoptera frugiperda,Sf 21 (baculovirus)-derived human Contactin-1 protein, (N-terminus) Human Contactin-1 (Glu21-Ser993) Accession &Num; CAA79696 IEGRMD Human IgG1 (Pro100-Lys330) (C-terminus) |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. |
| Structural Form | Disulfide-linked homodimer |
| Conjugate | Unconjugated |
| Recombinant | Recombinant |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?