missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Human Complement Component C1s Protein
Description
The Recombinant Human Complement Component C1s Protein is derived from NS0. The Recombinant Human Complement Component C1s Protein has been validated for the following applications: Enzyme Activity.
Specifications
Specifications
| Formulation | Supplied as a 0.2 μm filtered solution in Tris, NaCl, CaCl 2 and Glycerol. |
| Gene ID (Entrez) | 716 |
| Molecular Weight (g/mol) | 47 kDa (A chain) & 28 kDa (B chain) |
| Name | Complement Component C1s |
| Quantity | 10 μg |
| Source | Mouse myeloma cell line,NS0-derived human Complement Component C1s protein Met1-Asp688,with a C-terminal 6-His tag |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -20 to -70° C as supplied. 3 months, -20 to -70° C under sterile conditions after opening. |
| Structural Form | Disulfide-linked heterodimer |
| Conjugate | Unconjugated |
| Recombinant | Recombinant |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?