Learn More
DiscoverX Recombinant Human Complement C5a
Product Code. 17312439
Encompass
Click to view available options
:
25 μg
Product Code. 17312439
Brand: DiscoverX 921103
Recombinant Human Complement C5a is an agonist for C5AR1 and C5AR2. Provided as 25μg of dry powder alongside reconstitution buffer.
Complement Component 5a (C5a) is a pro-inflammatory protein fragment anaphylatoxin, and is cleaved by C5-convertase from complement component C5. Acting as an agonist for C5AR1 and C5AR2 on macrophages, neutrophils, and endothelial cells, C5a is a strong chemoattractant in stimulating an inflammatory response, and is recruited with the complement system network during an immune response to tissue damage or foreign objects. It is widely involved in many forms of inflammatory diseases, including asthma, ischemia-reperfusion injury, acute lung injury, and sepsis. Provided as 25μg of dry powder alongside reconstitution buffer.
Specifications
Quantity 25 μg
Product Type Control Ligands