missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Human CNTF Protein

Product Code. p-4719365
Click to view available options
Quantity:
10 μg
50 μg
Formulation:
Lyophilized from a 0.2μm filtered solution in PBS, NaCl, EDTA, DTT and CHAPS with BSA as a carrier protein.
Lyophilized from a 0.2μm filtered solution in PBS, NaCl, EDTA, DTT and CHAPS.
Unit Size:
10µg
50µg
This item is not returnable. View return policy

Product Code. 18178520

Brand: R&D Systems™ 257NT010

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity

The Recombinant Human CNTF Protein is derived from E. coli. The Recombinant Human CNTF Protein has been validated for the following applications: Bioactivity.
TRUSTED_SUSTAINABILITY

Spezifikation

Formulation Lyophilized from a 0.2μm filtered solution in PBS, NaCl, EDTA, DTT and CHAPS with BSA as a carrier protein.
Gene ID (Entrez) 1270
Molecular Weight (g/mol) 22.8 kDa
Name CNTF
Quantity 10 μg
Source E. coli-derived human CNTF protein Ala2-Met200
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 3 months, 2 to 8° C under sterile conditions after reconstitution.
Structural Form This recombinant protein is prone to proteolytic degradation at the C-terminus, resulting in proteins with molecular masses of 22.2 - 22.8 kDa.
Endotoxin Concentration <0.1 EU per 1μg of the protein by the LAL method.
Conjugate Unconjugated
Recombinant Recombinant
Activity Measured in a cell proliferation assay using TF-1 human erythroleukemic cells. Kitamura, T. et al. (1989) J. Cell Physiol. 140:323. The ED50 for this effect is 0.03-0.18 μg/mL. The proliferative effect can be enhanced up to 100-fold by the addition of Recombinant Human CNTF R alpha (Catalog # 303-CR ) .
Purity or Quality Grade 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Mehr anzeigen Weniger anzeigen
Berichtigung von Produktinhalten

Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.

Name des Produkts

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.

Vielen Dank, dass Sie uns helfen, unsere Website zu verbessern. Ihr Feedback wurde übermittelt