missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Human CNTF Protein
Description
The Recombinant Human CNTF Protein is derived from E. coli. The Recombinant Human CNTF Protein has been validated for the following applications: Bioactivity.
Specifications
Specifications
| Formulation | Lyophilized from a 0.2μm filtered solution in PBS, NaCl, EDTA, DTT and CHAPS with BSA as a carrier protein. |
| Gene ID (Entrez) | 1270 |
| Molecular Weight (g/mol) | 22.8 kDa |
| Name | CNTF |
| Quantity | 10 μg |
| Source | E. coli-derived human CNTF protein Ala2-Met200 |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 3 months, 2 to 8° C under sterile conditions after reconstitution. |
| Structural Form | This recombinant protein is prone to proteolytic degradation at the C-terminus, resulting in proteins with molecular masses of 22.2 - 22.8 kDa. |
| Endotoxin Concentration | <0.1 EU per 1μg of the protein by the LAL method. |
| Conjugate | Unconjugated |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?