missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human CNDP2 / CPGL / PEPA Protein

Product Code. 16082101
Click to view available options
Quantity:
1 mg
10 μg
100 μg
200 μg
50 μg
500 μg
Unit Size:
100µg
10µg
1mg
200µg
500µg
50µg
This item is not returnable. View return policy

Product Code. 16082101

Brand: enQuireBio™ QP8005ec100ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

A DNA sequence encoding the Homo sapiens (Human) CNDP2 / CPGL / PEPA, was expressed in the hosts and tags indicated.

Specifications

Accession Number NP_001161971.1
Gene ID (Entrez) 55748
Name CNDP2 / CPGL / PEPA Protein
Quantity 100 μg
Regulatory Status Research Use Only
Endotoxin Concentration Not determined.
Gene Symbol CNDP2
Protein Length Full Length
Product Type Recombinant Protein
Cross Reactivity Human
Species E. coli
Protein Tag His
Sequence AALTTLFKYI DENQDRYIKK LAKWVAIQSV SAWPEKRGEI RRMMEVAAAD VKQLGGSVEL VDIGKQKLPD GSEIPLPPIL LGRLGSDPQK KTVCIYGHLD VQPAALEDGW DSEPFTLVER DGKLYGRGST DDKGPVAGWI NALEAYQKTG QEIPVNVRFC LEGMEESGSE GLDELIFARK DTFFKDVDYV CISDNYWLGK KKPCITYGLR GICYFFIEVE CSNKDLHSGV YGGSVHEAMT DLILLMGSLV DKRGNILIPG INEAVAAVTE EEHKLYDDID FDIEEFAKDV GAQILLHSHK KDILMHRWRY PSLSLHGIEG AFSGSGAKTV IPRKVVGKFS IRLVPNMTPE VVGEQVTSYL TKKFAELRSP NEFKVYMGHG GKPWVSDFSH PHYLAGRRAM KTVFGVEPDL TREGGSIPVT LTFQEATGKN VMLLPVGSAD DGAHSQNEKL NRYNYIEGTK MLAAYLYEVS QLKD
Buffer Tris-based buffer, 50% glycerol
Purity or Quality Grade Greater than 90% as determined by SDS-PAGE.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.