missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human CMC1 Protein

A cDNA sequence encoding the CMC1 was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP11457-5ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15937328

  • £99.15 / 5µg
Estimated Shipment: 12-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

CMC1 Protein
Research Use Only
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MGSMALDPAD QHLRHVEKDV LIPKIMREKA KERCSEQVQD FTKCCKNSGV LMVVKCRKEN SALKECLTAY YNDPAFYEEC KMEYLKEREE FRKTGIPTKK RLQKLPTSM
Greater than 85.0% as determined by SDS-PAGE.
5 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
His
CMC1 protein solution (0.5 mg/ml) contains 20mM Tris-HCl buffer, (pH 8.0), 0.4M UREA and 10% glycerol.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers