missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human CMA1 / Chymase 1 Protein
A DNA sequence encoding the Homo sapiens (Human) CMA1 / Chymase 1, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP5844-ec-500ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001295012.1 | |
CMA1 / Chymase 1 Protein | |
Research Use Only | |
CMA1 | |
Recombinant Protein | |
E. coli | |
IIGGTECKPH SRPYMAYLEI VTSNGPSKFC GGFLIRRNFV LTAAHCAGRS ITVTLGAHNI TEEEDTWQKL EVIKQFRHPK YNTSTLHHDI MLLKLKEKAS LTLAVGTLPF PSQFNFVPPG RMCRVAGWGR TGVLKPGSDT LQEVKLRLMD PQACSHFRDF DHNLQLCVGN PRKTKSAFKG DSGGPLLCAG VAQGIVSYGR SDAKPPAVFT RISHYRPWIN QILQAN | |
Greater than 90% as determined by SDS-PAGE. |
1215 | |
500 μg | |
Not determined. | |
Full Length | |
Human | |
His | |
Tris-based buffer, 50% glycerol |