missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Human CLF-1/CLC Complex Protein
Click to view available options
Formulation:
Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein.
Lyophilized from a 0.2μm filtered solution in PBS.
Unit Size:
25µg
Description
The Recombinant Human CLF-1/CLC Complex Protein is derived from NS0. The Recombinant Human CLF-1/CLC Complex Protein has been validated for the following applications: Bioactivity.
Specifications
Specifications
| Formulation | Lyophilized from a 0.2μm filtered solution in PBS. |
| Gene ID (Entrez) | 9244 |
| Molecular Weight (g/mol) | 44.5 kDa (CLF-1) & 23 kDa (CLC) |
| Name | CLF-1/CLC Complex |
| Quantity | 25 μg |
| Source | Mouse myeloma cell line,NS0-derived human CLF-1/CLC Complex protein, (N-terminus) Human CLF-1 (Ala38-Arg422) Accession &Num;O75462.1 10-His tag Human CLC (Leu28-Phe225) Accession &Num; Q9UBD9 6-His tag (C-terminus) |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. |
| Conjugate | Unconjugated |
| Recombinant | Recombinant |
| Purity or Quality Grade | 85%, by SDS-PAGE under reducing conditions and visualized by silver stain. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction