missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human CLEC2B Protein
A cDNA sequence encoding the CLEC2B was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP11444-5ug
This item is not returnable.
View return policy
Specifications
CLEC2B Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSKLTRDSQ SLCPYDWIGF QNKCYYFSKE EGDWNSSKYN CSTQHADLTI IDNIEEMNFL RRYKCSSDHW IGLKMAKNRT GQWVDGATFT KSFGMRGSEG CAYLSDDGAA TARCYTERKW ICRKRIH | |
Greater than 90.0% as determined by SDS-PAGE. |
5 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
CLEC2B protein solution (1 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0), 2M UREA and 10% glycerol. |