missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Clathrin interactor 1 Protein

Product Code. 15978139
Click to view available options
Quantity:
1 mg
10 μg
100 μg
200 μg
50 μg
500 μg
Unit Size:
100µg
10µg
1mg
200µg
500µg
50µg
This item is not returnable. View return policy

Product Code. 15978139

Brand: enQuireBio™ QP5839ye100ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

A DNA sequence encoding the Homo sapiens (Human) Clathrin interactor 1, was expressed in the hosts and tags indicated.

Specifications

Accession Number NP_001182484.1
Gene ID (Entrez) 9685
Name Clathrin interactor 1 Protein
Quantity 100 μg
Regulatory Status Research Use Only
Endotoxin Concentration Not determined.
Gene Symbol CLINT1
Protein Length Full Length
Product Type Recombinant Protein
Cross Reactivity Human
Species Yeast
Protein Tag His
Sequence MLNMWKVREL VDKATNVVMN YSEIESKVRE ATNDDPWGPS GQLMGEIAKA TFMYEQFPEL MNMLWSRMLK DNKKNWRRVY KSLLLLAYLI RNGSERVVTS AREHIYDLRS LENYHFVDEH GKDQGINIRQ KVKELVEFAQ DDDRLREERK KAKKNKDKYV GVSSDSVGGF RYSERYDPEP KSKWDEEWDK NKSAFPFSDK LGELSDKIGS TIDDTISKFR RKDREDSPER CSDSDEEKKA RRGRSPKGEF KDEEETVTTK HIHITQATET TTTRHKRTAN PSKTIDLGAA AHYTGDKASP DQNASTHTPQ SSVKTSVPSS KSSGDLVDLF DGTSQSTGGS ADLFGGFADF GSAAASGSFP SQVTATSGNG DFGDWSAFNQ APSGPVASSG EFFGSASQPA VELVSGSQSA LGPPPAASNS SDLFDLMGSS QATMTSSQSM NFSMMSTNTV GLGLPMSRSQ NTDMVQKSVS KTLPSTWSDP SVNISLDNLL PGMQPSKPQQ PSLNTMIQQQ NMQQPMNVMT QSFGAVNLSS PSNMLPVRPQ TNALIGGPMP MSMPNVMTGT MGMAPLGNTP MMNQSMMGMN MNIGMSAAGM GLTGTMGMGM PNIAMTSGTV QPKQDAFANF ANFSK
Buffer Tris-based buffer, 50% glycerol
Purity or Quality Grade Greater than 90% as determined by SDS-PAGE.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.