missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Clathrin heavy chain 2 Protein

A DNA sequence encoding the Homo sapiens (Human) Clathrin heavy chain 2, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP5842-ye-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15948169

  • £467.00 / 100µg
Estimated Shipment: 20-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_001826.3
Clathrin heavy chain 2 Protein
Research Use Only
CLTCL1
Recombinant Protein
Yeast
LLVLSPRLDH TWTVSFFSKA GQLPLVKPYL RSVQSHNNKS VNEALNHLLT EEEDYQGLRA SIDAYDNFDN ISLAQQLEKH QLMEFRCIAA YLYKGNNWWA QSVELCKKDH LYKDAMQHAA ESRDAELAQK LLQWFLEEGK RECF
Greater than 90% as determined by SDS-PAGE.
8218
100 μg
Not determined.
Partial (see sequence information for more details).
Human
His
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers