missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human CKS2 Protein
A cDNA sequence encoding the CKS2 was constructed and used to recombinantly synthesize the protein.
£99.15 - £2975.00
Specifications
Name | CKS2 Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Product Type | Recombinant Protein |
Cross Reactivity | Human |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15957268
|
enQuireBio™
QP11439-5UG |
5 μg |
£99.15
5µg |
Estimated Shipment: 07-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15947268
|
enQuireBio™
QP11439-25UG |
25 μg |
£155.00
25µg |
Estimated Shipment: 07-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15937268
|
enQuireBio™
QP11439-1MG |
1 mg |
£2975.00
1mg |
Estimated Shipment: 07-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
CKS2 Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
Untagged | |
The CKS2 protein solution contains 20mM Tris-HCl pH 7.5 and 20% glycerol. |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MASMTGGQQM GRGSHMAHKQ IYYSDKYFDE HYEYRHVMLP RELSKQVPKT HLMSEEEWRRLGVQQSLGWV HYMIHEPEPH ILLFRRPLPK DQQ | |
Greater than 95.0% as determined by SDS-PAGE. |