missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human CISD1 Protein
A cDNA sequence encoding the CISD1 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP11430-1mg
Additional Details : Weight : 0.01000kg
Specifications
CISD1 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSKRFYVKD HRNKAMINLH IQKDNPKIVH AFDMEDLGDK AVYCRCWRSK KFPFCDGAHT KHNEETGDNV GPLIIKKKET | |
Greater than 90% as determined by SDS-PAGE. |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
The CISD1 solution contains 20mM Tris-HCl buffer (pH 8.0), 0.15M NaCl, 1mM DTT and 10% glycerol. |