missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Human cIAP-2 (HIAP-1) Protein
Click to view available options
Quantity:
50 μg
Unit Size:
50µg
Description
The Recombinant Human cIAP-2 (HIAP-1) Protein is derived from E. coli. The Recombinant Human cIAP-2 (HIAP-1) Protein has been validated for the following applications: Inhibition Activity.
Specifications
Specifications
| Formulation | Supplied as a 0.2μm filtered solution in HEPES, NaCl, DTT and Glycerol. |
| Gene ID (Entrez) | 330 |
| Molecular Weight (g/mol) | 71 kDa |
| Name | cIAP-2/HIAP-1 |
| Quantity | 50 μg |
| Source | E. coli-derived human cIAP-2/HIAP-1 protein, (N-terminus) ATVID 10-His tag SIEGRA Human cIAP-2 (Asn2-Ser604) Accession &Num; Q13489 (C-terminus) |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -70° C as supplied. 3 months, -70° C under sterile conditions after opening. |
| Conjugate | Unconjugated |
| Recombinant | Recombinant |
| Purity or Quality Grade | 95%, by SDS-PAGE under reducing conditions and visualized by silver stain. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction