missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Chymotrypsin-like elastase family member 2A Protein
A DNA sequence encoding the Homo sapiens (Human) Chymotrypsin-like elastase family member 2A, was expressed in the hosts and tags indicated.
Marque: enQuireBio™ QP5816-ec-10ug
Informations supplémentaires : Weight : 0.01000kg
Spécification
NP_254275.1 | |
Chymotrypsin-like elastase family member 2A Protein | |
Research Use Only | |
Cela2a | |
Recombinant Protein | |
E. coli | |
VVGGEEARPN SWPWQVSLQY SSNGKWYHTC GGSLIANSWV LTAAHCISSS RTYRVGLGRH NLYVAESGSL AVSVSKIVVH KDWNSNQISK GNDIALLKLA NPVSLTDKIQ LACLPPAGTI LPNNYPCYVT GWGRLQTNGA VPDVLQQGRL LVVDYATCSS SAWWGSSVKT SMICAGGDGV ISSCNGDSGG PLNCQASDGR WQVHGIVSFG SRLGCNYYHK PSVFTRVSNY IDWINSVIAN N | |
Greater than 80% as determined by SDS-PAGE. |
63036 | |
10 μg | |
Not determined. | |
Full Length | |
Human | |
His | |
Tris-based buffer, 50% glycerol |