missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human CHIC2 Protein
A DNA sequence encoding the Homo sapiens (Human) CHIC2, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8255-ec-500ug
Additional Details : Weight : 0.01000kg
Specifications
NP_036242.1 | |
CHIC2 Protein | |
Research Use Only | |
CHIC2 | |
Recombinant Protein | |
E. coli | |
MADFDEIYEE EEDEERALEE QLLKYSPDPV VVRGSGHVTV FGLSNKFESE FPSSLTGKVA PEEFKASINR VNSCLKKNLP VNVRWLLCGC LCCCCTLGCS MWPVICLSKR TRRSIEKLLE WENNRLYHKL CLHWRLSKRK CETNNMMEYV ILIEFLPKTP IFRPD | |
Greater than 80% as determined by SDS-PAGE. |
26511 | |
500 μg | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |