missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human CHIC2 Protein

A DNA sequence encoding the Homo sapiens (Human) CHIC2, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP8255-ec-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16083591

  • £2480.00 / 1mg
Estimated Shipment: 10-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_036242.1
CHIC2 Protein
Research Use Only
CHIC2
Recombinant Protein
E. coli
MADFDEIYEE EEDEERALEE QLLKYSPDPV VVRGSGHVTV FGLSNKFESE FPSSLTGKVA PEEFKASINR VNSCLKKNLP VNVRWLLCGC LCCCCTLGCS MWPVICLSKR TRRSIEKLLE WENNRLYHKL CLHWRLSKRK CETNNMMEYV ILIEFLPKTP IFRPD
Greater than 80% as determined by SDS-PAGE.
26511
1 mg
Not determined.
Full Length
Human
GST
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers