missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Charged multivesicular body protein 2a Protein

A DNA sequence encoding the Homo sapiens (Human) Charged multivesicular body protein 2a, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP8478-ec-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16004841

  • £1735.00 / 1mg
Estimated Shipment: 14-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_055268.1
Charged multivesicular body protein 2a Protein
Research Use Only
BC2
Recombinant Protein
E. coli
MDLLFGRRKT PEELLRQNQR ALNRAMRELD RERQKLETQE KKIIADIKKM AKQGQMDAVR IMAKDLVRTR RYVRKFVLMR ANIQAVSLKI QTLKSNNSMA QAMKGVTKAM GTMNRQLKLP QIQKIMMEFE RQAEIMDMKE EMMNDAIDDA MGDEEDEEES DAVVSQVLDE LGLSLTDELS NLPSTGGSLS VAAGGKKAEA AASALADADA DLEERLKNLR RD
Greater than 90% as determined by SDS-PAGE.
27243
1 mg
Not determined.
Full Length
Human
GST
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers