missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Cerebellar degeneration-related protein 1
A DNA sequence encoding the Homo sapiens (Human) Cerebellar degeneration-related protein 2, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP5810-ec-50ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001793.1 | |
Cerebellar degeneration-related protein 1 | |
Research Use Only | |
CDR2 | |
Recombinant Protein | |
E. coli | |
MLAENLVEEF EMKEDEPWYD HQDLQQDLQL AAELGKTLLD RNTELEDSVQ QMYTTNQEQL QEIEYLTKQV ELLRQMNEQH AKVYEQLDVT ARELEETNQK LVADSKASQQ KILSLTETIE CLQTNIDHLQ SQVEELKSSG QGRRSPGKCD QEKPAPSFAC LKELYDLRQH FVYDHVFAEK ITSLQGQPSP DEEENEHLKK TVTMLQAQLS LERQKRVTME EEYGLVLKEN SELEQQLGAT GAYRARALEL EAEVAEMRQM LQSEHPFVNG VEKLVPDSLY VPFKEPSQSL LEEMFLTVPE SHRKPLKRSS SETILSSLAG SDIVKGHEET CIRRAKAVKQ RGISLLHEVD TQYSALKVKY EELLKKCQEE QDSLSHKAVQ TSRAAAKDLT GVNAQSEPVA SGWELASVNP EPVSSPTTPP EYKALFKEIF SCIKKTKQEI DEQRTKYRSL SSHS | |
Greater than 90% as determined by SDS-PAGE. |
101060399 | |
50 μg | |
Not determined. | |
Full Length | |
Human | |
His | |
Tris-based buffer, 50% glycerol |