missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Cerebellar degeneration-related protein 1

A DNA sequence encoding the Homo sapiens (Human) Cerebellar degeneration-related protein 2, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP5810-ec-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15927919

  • £384.00 / 100µg
Estimated Shipment: 14-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_001793.1
Cerebellar degeneration-related protein 1
Research Use Only
CDR2
Recombinant Protein
E. coli
MLAENLVEEF EMKEDEPWYD HQDLQQDLQL AAELGKTLLD RNTELEDSVQ QMYTTNQEQL QEIEYLTKQV ELLRQMNEQH AKVYEQLDVT ARELEETNQK LVADSKASQQ KILSLTETIE CLQTNIDHLQ SQVEELKSSG QGRRSPGKCD QEKPAPSFAC LKELYDLRQH FVYDHVFAEK ITSLQGQPSP DEEENEHLKK TVTMLQAQLS LERQKRVTME EEYGLVLKEN SELEQQLGAT GAYRARALEL EAEVAEMRQM LQSEHPFVNG VEKLVPDSLY VPFKEPSQSL LEEMFLTVPE SHRKPLKRSS SETILSSLAG SDIVKGHEET CIRRAKAVKQ RGISLLHEVD TQYSALKVKY EELLKKCQEE QDSLSHKAVQ TSRAAAKDLT GVNAQSEPVA SGWELASVNP EPVSSPTTPP EYKALFKEIF SCIKKTKQEI DEQRTKYRSL SSHS
Greater than 90% as determined by SDS-PAGE.
101060399
100 μg
Not determined.
Full Length
Human
His
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers