missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human CENPQ Protein
A cDNA sequence encoding the CENPQ was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP11391-1mg
Additional Details : Weight : 0.01000kg
Specifications
CENPQ Protein | |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
CENPQ protein solution (1 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0) and 10% glycerol. |
Greater than 85.0% as determined by SDS-PAGE. | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMSGKANA SKKNAQQLKR NPKRKKDNEE VVLSENKVRN TVKKNKNHLK DLSSEGQTKH TNLKHGKTAA SKRKTWQPLS KSTRDHLQTM MESVIMTILS NSIKEKEEIQ YHLNFLKKRL LQQCETLKVP PKKMEDLTNV SSLLNMERAR DKANEEGLAL LQEEIDKMVE TTELMTGNIQ SLKNKIQILA SEVEEEEERV KQMHQINSSG VLSLPELSQK TLKAPTLQKE ILALIPNQNA LLKDLDILHN SSQMKSMSTF IEEAYKKLDA S |